Invitrogen
Manufacturer Code:PA5100664
Catalog # PIPA5100664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GNA13 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU024132 AU043124 G alpha 13 G alpha-13 G protein subunit alpha 13 G13 Galpha 13 Galpha13 Gna13 Gna-13 G-protein subunit alpha-13 guanine nucleotide binding protein (G protein) alpha 13 guanine nucleotide binding protein alpha 13 guanine nucleotide-binding protein subunit alpha-13; G alpha-13; G-protein subunit alpha-13; guanine nucleotide binding protein (G protein), alpha 13; guanine nucleotide binding protein, alpha 13; Guanine nucleotide-binding protein subunit alpha-13
Gene Aliases: AU024132; AU043124; G13; Galpha13; Gna-13; GNA13
UniProt ID: (Human) F5H1G8, (Mouse) P27601, (Rat) Q6Q7Y5
Entrez Gene ID: (Human) 10672, (Mouse) 14674, (Rat) 303634
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.