Invitrogen
Manufacturer Code:PA529928
Catalog # PIPA529928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 162 and 381 of Human G protein alpha 12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G alpha-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; Guanine nucleotide-binding protein subunit alpha-12; NNX3 RMP gep G-protein subunit alpha-12 WUGSC:H&lowbarGS165O14.2 g alpha-12 guanine nucleotide-binding protein subunit alpha-12; WUGSC:H_GS165O14.2
Gene Aliases: gep; GNA12; NNX3; RMP
UniProt ID: (Human) Q03113
Entrez Gene ID: (Human) 2768
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.