Invitrogen
Manufacturer Code:PA5100663
Catalog # PIPA5100663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GNA12 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI414047 AI504261 G alpha-12 G protein subunit alpha 12 Galpha12 gep GNA12 Gna-12 G-protein subunit alpha-12 guanine nucleotide binding protein (G protein) alpha 12 guanine nucleotide binding protein alpha 12 guanine nucleotide-binding protein subunit alpha-12 NNX3 RMP WUGSC:H_GS165O14.2; G alpha-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; guanine nucleotide binding protein, alpha 12; Guanine nucleotide-binding protein subunit alpha-12; WUGSC:H_GS165O14.2
Gene Aliases: AI414047; AI504261; Galpha12; gep; Gna-12; GNA12; NNX3; RMP
UniProt ID: (Human) Q03113, (Mouse) P27600, (Rat) Q63210
Entrez Gene ID: (Human) 2768, (Mouse) 14673, (Rat) 81663
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.