Abnova
Manufacturer Code:H00002768A01
Catalog # 89-104-555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 98 a.a) full-length recombinant protein with GST tag.,GNA12 (AAH09457 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Protein Aliases: G alpha-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; Guanine nucleotide-binding protein subunit alpha-12; WUGSC:H_GS165O14.2
Gene Aliases: gep; GNA12; NNX3; RMP
UniProt ID: (Human) Q03113
Entrez Gene ID: (Human) 2768
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.