Invitrogen
Manufacturer Code:PA570664
Catalog # PIPA570664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human GNA11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G alpha-11; G-protein subunit alpha-11; GNA-11 G-protein subunit alpha-11 g alpha-11 guanine nucleotide-binding protein G(y) subunit alpha guanine nucleotide-binding protein subunit alpha-11 guanine nucleotide-binding protein Gq class GNA11; guanine nucleotide binding protein (G protein), alpha 11 (Gq class); guanine nucleotide regulatory protein G alpha 11; guanine nucleotide-binding protein alpha 11; guanine nucleotide-binding protein alpha 11 subunit; Guanine nucleotide-binding protein G(y) subunit alpha; Guanine nucleotide-binding protein subunit alpha-11
Gene Aliases: Dsk7; E430025L19; FBH; FBH2; FHH2; GA11; GNA-11; GNA11; HHC2; HYPOC2
UniProt ID: (Human) P29992, (Mouse) P21278, (Rat) Q9JID2
Entrez Gene ID: (Goat) 102177328, (Horse) 100067327, (Human) 2767, (Sheep) 101119215, (Mouse) 14672, (Guinea pig) 100715351, (Rat) 81662
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.