Novus Biologicals
Manufacturer Code:NBP256670
Catalog # NBP256670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AGGDLKDGHHHYEGAVVILDAGAQYGKVIDRRVRELFVQSEIFPLETPAFAIKEQGFRAIIISGGPNSVYAEDAPWFDPAIFTIGKPVLGICYGMQMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.3.5.2 Glutamine amidotransferase GMP synthase GMP synthase [glutamine-hydrolyzing] GMP synthetase guanine monphosphate synthetase guanosine 5'-monophosphate synthase MLL/GMPS fusion protein; Glutamine amidotransferase; GMP synthase; GMP synthase [glutamine-hydrolyzing]; GMP synthetase; guanine monophosphate synthetase; guanosine 5'-monophosphate synthase; MLL/GMPS fusion protein; testicular tissue protein Li 82
Gene Aliases: GMPS
UniProt ID: (Human) P49915
Entrez Gene ID: (Human) 8833
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.