Invitrogen
Manufacturer Code:PA595784
Catalog # PIPA595784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit funsion protein containing a sequence corresponding to amino acids 50-250 of human GMPPB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDP-mannose pyrophosphorylase B; GTP-mannose-1-phosphate guanylyltransferase beta; Mannose-1-phosphate guanyltransferase beta
Gene Aliases: AI317178; E430010H19; Gmppb; RGD1560458
UniProt ID: (Mouse) Q8BTZ7
Entrez Gene ID: (Rat) 363145, (Mouse) 331026
Molecular Function: G-protein modulator RNA binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding nucleotidyltransferase transferase translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.