Proteintech
Manufacturer Code:136251AP150UL
Catalog # 50-172-8886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GMFG Fusion Protein Ag4538 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glia maturation factor gamma; Glia maturation factor gamma glia maturation factor gamma GMF GAMMA GMFG; glia maturation factor, gamma; GMF-gamma
Gene Aliases: 0610039G16Rik; 2310057N07Rik; AI324845; GMF-GAMMA; GMFG
UniProt ID: (Human) O60234, (Rat) Q80T18, (Mouse) Q9ERL7
Entrez Gene ID: (Human) 9535, (Rat) 113940, (Mouse) 63986
Molecular Function:
enzyme modulator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.