Proteintech
Manufacturer Code:106901AP150UL
Catalog # 50-172-6517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GMF-beta Fusion Protein Ag1104 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA for thyroid hormone receptor binding site (276bp); Glia maturation factor beta; Glia maturation factor beta glia maturation factor beta GMF GMF beta GMFB GMF-beta; glia maturation factor, beta; GMF-beta
Gene Aliases: 3110001H22Rik; 3110001O16Rik; AI851627; C79176; D14Ertd630e; GMF; GMFB
UniProt ID: (Human) P60983, (Rat) Q63228, (Mouse) Q9CQI3
Entrez Gene ID: (Human) 2764, (Rat) 81661, (Mouse) 63985
Molecular Function: enzyme modulator kinase modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.