Novus Biologicals
Manufacturer Code:NBP270835AF700
Catalog # N270835A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A1 |
Immunogen | Full length recombinant protein of human GMDS (NP_001491) produced in HEK293T cells. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 4.2.1.47 GDP-D-mannose dehydratase GDP-mannose 46-dehydratase GMDGDP-mannose 46 dehydratase SDR3E1 short chain dehydrogenase/reductase family 3E member 1; GDP-D-mannose dehydratase; GDP-mannose 4,6 dehydratase; GMD; short chain dehydrogenase/reductase family 3E, member 1
Gene Aliases: GMD; GMDS; SDR3E1
UniProt ID: (Human) O60547
Entrez Gene ID: (Human) 2762
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.