Invitrogen
Manufacturer Code:PA568194
Catalog # PIPA568194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human GMDS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDP-D-mannose dehydratase; GDP-mannose 4,6 dehydratase; GMD; RP1-118B18.1 GMD SDR3E1 GDP-D-mannose dehydratase GDP-mannose 46 dehydratase short chain dehydrogenase/reductase family 3E member 1; short chain dehydrogenase/reductase family 3E, member 1
Gene Aliases: BC031788; C87208; GMD; GMDS; SDR3E1
UniProt ID: (Human) O60547, (Mouse) Q8K0C9
Entrez Gene ID: (Human) 2762, (Mouse) 218138, (Rat) 291095
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.