Invitrogen
Manufacturer Code:PA556771
Catalog # PIPA556771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GMDS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDP-D-mannose dehydratase; GDP-mannose 4,6 dehydratase; GMD; RP1-118B18.1 GMD SDR3E1 GDP-D-mannose dehydratase GDP-mannose 46 dehydratase short chain dehydrogenase/reductase family 3E member 1; short chain dehydrogenase/reductase family 3E, member 1
Gene Aliases: GMD; GMDS; SDR3E1
UniProt ID: (Human) O60547
Entrez Gene ID: (Human) 2762
Molecular Function:
dehydratase
epimerase/racemase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.