Invitrogen
Manufacturer Code:PA532783
Catalog # PIPA532783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from N-terminus of human GM2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerebroside sulfate activator protein; Ganglioside GM2 activator; Ganglioside GM2 activator isoform short; GM2-AP; GM2-AP SAP-3 cerebroside sulfate activator protein ganglioside GM2 activator shingolipid activator protein 3 sphingolipid activator protein 3; SAP-3; shingolipid activator protein 3; Sphingolipid activator protein 3
Gene Aliases: GM2-AP; GM2A; SAP-3
UniProt ID: (Human) P17900
Entrez Gene ID: (Human) 2760
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.