Novus Biologicals
Manufacturer Code:NBP300106AF647
Catalog # NB086737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant GM2A (AAA35907.1 Met1-Ile193). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerebroside sulfate activator protein; Cerebroside sulfate activator protein ganglioside GM2 activator GM2 ganglioside activator GM2-AP SAP-3GM2 ganglioside activator protein Shingolipid activator protein 3 sphingolipid activator protein 3; Ganglioside GM2 activator; Ganglioside GM2 activator isoform short; GM2-AP; SAP-3; shingolipid activator protein 3; Sphingolipid activator protein 3
Gene Aliases: GM2-AP; GM2A; SAP-3
UniProt ID: (Human) P17900
Entrez Gene ID: (Human) 2760
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.