Proteintech
Manufacturer Code:108642AP150UL
Catalog # 50-172-6667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GM2A Fusion Protein Ag1295 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerebroside sulfate activator protein; Ganglioside GM2 activator; Ganglioside GM2 activator GM2 AP GM2 ganglioside activator GM2A SAP 3; Ganglioside GM2 activator isoform short; GM2 ganglioside activator protein; GM2-AP; SAP-3; shingolipid activator protein 3; Sphingolipid activator protein 3
Gene Aliases: AA408702; AW215435; GM2-AP; GM2A; SAP-3
UniProt ID: (Human) P17900, (Mouse) Q60648
Entrez Gene ID: (Human) 2760, (Mouse) 14667, (Rat) 282838
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.