Invitrogen
Manufacturer Code:PA1077A555
Catalog # PIPA1077555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues G(368) Q V M E S V R Q L Q M E R D K(383) of human GM130 |
Conjugate | Alexa Fluor 555 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa cis-Golgi matrix protein; GM130; GM130 autoantigen; GM130 RP11-395P17.5; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; Golgin subfamily A member 2; Golgin-95; SY11 protein
Gene Aliases: GM130; GOLGA2
UniProt ID: (Human) Q08379
Entrez Gene ID: (Human) 2801
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.