Invitrogen
Manufacturer Code:PA547982
Catalog # PIPA547982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit feline GM-CSF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Colony-stimulating factor; CSF; GM-CSF; Granulocyte-macrophage colony-stimulating factor; Granulocyte-macrophage colony-stimulating factor GM-CSF Colony-stimulating factor CSF Molgramostin Sargramostim CSF2 GMCSF
Gene Aliases: CSF2; GM-CSF
UniProt ID: (Cat) O62757
Entrez Gene ID: (Cat) 493805
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.