R&D Systems
Manufacturer Code:AF1546
Catalog # AF1546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant canine GM-CSF Ala18-Lys144 Accession # P48749.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: colony stimulating factor 2 (granulocyte-macrophage); colony stimulating factor 2 (granulocyte-macrophage) Colony-stimulating factor CSF CSF2 CSF-2 GMCSF GM-CSF GMCSFgranulocyte-macrophage colony-stimulating factor granulocyte-macrophage colony stimulating factor MGC131935 MGC138897 Molgramostim; Colony-stimulating factor; CSF; GM-CSF; granulocyte macrophage-colony stimulating factor; Granulocyte-macrophage colony-stimulating factor; Molgramostin; Sargramostim
Gene Aliases: CSF2; GMCSF
UniProt ID: (Human) P04141
Entrez Gene ID: (Human) 1437
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.