Proteintech
Manufacturer Code:148331AP150UL
Catalog # 50-172-9763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLYR1 Fusion Protein Ag6637 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyisobutyrate dehydrogenase-like protein; BM045 Glyoxylate reductase 1 homolog GLYR1 HIBDL N PAC N-PAC Nuclear protein NP60 Nuclear protein of 60 kDa Putative oxidoreductase GLYR1; Cytokine-like nuclear factor N-PAC; Glyoxylate reductase 1 homolog; hNDF; nuclear protein 60 kDa; nuclear protein 60kDa; Nuclear protein NP60; Nuclear protein of 60 kDa; Nucleosome-destabilizing factor; Putative oxidoreductase GLYR1
Gene Aliases: 2810419J22Rik; 3930401K13Rik; AW545332; BM045; GLYR1; HIBDL; N-PAC; NDF; NP60; Npac; RGD1309459
UniProt ID: (Human) Q9BTI1, (Rat) Q5RKH0, (Mouse) Q922P9
Entrez Gene ID: (Human) 84656, (Rat) 360477, (Mouse) 74022
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.