Invitrogen
Manufacturer Code:PA569588
Catalog # PIPA569588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human GLYATL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:glycine-N-acyltransferase-like protein 3; Glycine N-acyltransferase-like protein 3; glycine N-acyltransferase-like protein 3-like; glycine-N-acyltransferase-like 3; Glycine-N-acyltransferase-like protein 3
Gene Aliases: bA28H17.2; C6orf140; EG435528; GLYATL3; Gm5683
UniProt ID: (Human) Q5SZD4, (Mouse) E9Q5L8
Entrez Gene ID: (Bovine) 787783, (Human) 389396, (Dog) 610094, (Guinea pig) 100726471, (Rat) 688536, (Mouse) 435528
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.