Novus Biologicals
Manufacturer Code:NBP17931420UL
Catalog # NBP17931420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human C6orf140. Peptide sequence NPFQKEVVLDSWPDFKAVITRRQREAETDNLDHYTNAYAVFYKDVRAYRQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:glycine-N-acyltransferase-like protein 3; Glycine N-acyltransferase-like protein 3; glycine-N-acyltransferase-like 3
Gene Aliases: bA28H17.2; C6orf140; GLYATL3
UniProt ID: (Human) Q5SZD4
Entrez Gene ID: (Human) 389396
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.