Invitrogen
Manufacturer Code:PA521805
Catalog # PIPA521805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 73 and 302 of GLYATL1 (Uniprot IDNo.Q969I3) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:glycine N-acyltransferase-like protein 1; GATF-C GNAT acyl-CoA:glycine N-acyltransferase-like protein 1 glycine N-acyltransferase-like protein 1; Glutamine N-acyltransferase; Glycine N-acyltransferase-like protein 1; glycine-N-acyltransferase-like 1
Gene Aliases: GATF-C; GLYATL1; GNAT
UniProt ID: (Human) Q969I3
Entrez Gene ID: (Human) 92292
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.