Novus Biologicals
Manufacturer Code:NBP132600
Catalog # NBP132600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human GLYATL1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:glycine N-acyltransferase-like protein 1; Acyl-CoA:glycine N-acyltransferase-like protein 1 EC 2.3.1.13 FLJ26507 FLJ34646 GATF-C glycine N-acyltransferase-like protein 1 glycine-N-acyltransferase-like 1 GNAT MGC15397 MGC15937; Glutamine N-acyltransferase; Glycine N-acyltransferase-like protein 1; glycine-N-acyltransferase-like 1
Gene Aliases: GATF-C; GLYATL1; GNAT
UniProt ID: (Human) Q969I3
Entrez Gene ID: (Human) 92292
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.