Proteintech
Manufacturer Code:157171AP150UL
Catalog # 50-173-0429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLYATL1 Fusion Protein Ag8392 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:glycine N-acyltransferase-like protein 1; GATF C GLYATL1 GNAT; Glutamine N-acyltransferase; Glycine N-acyltransferase-like protein 1; glycine-N-acyltransferase-like 1
Gene Aliases: GATF-C; GLYATL1; GNAT
UniProt ID: (Human) Q969I3
Entrez Gene ID: (Human) 92292
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.