Novus Biologicals
Manufacturer Code:NBP271564AF647
Catalog # N271564A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D1 |
Immunogen | Full length recombinant protein of human GLYAT (NP_005829) produced in E.coli. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AAc; ACGNATAAc Acyl-CoA:glycine N-acyltransferase Aralkyl acyl-CoA N-acyltransferase Aralkyl acyl-CoA:amino acid N-acyltransferase aralkyl-CoA N-acyltransferase CATEC 2.3.1.13 GATHRP-1(CLP) glycine N-acyltransferase glycine-N-acyltransferase; Acyl-CoA:glycine N-acyltransferase; Aralkyl acyl-CoA N-acyltransferase; Aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; Benzoyl-coenzyme A:glycine N-acyltransferase; Glycine N-acyltransferase; Glycine N-benzoyltransferase; HRP-1(CLP)
Gene Aliases: ACGNAT; CAT; GAT; GLYAT
UniProt ID: (Human) Q6IB77
Entrez Gene ID: (Human) 10249
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.