Invitrogen
Manufacturer Code:PA565261
Catalog # PIPA565261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GLYAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAc; Acyl-CoA:glycine N-acyltransferase; Aralkyl acyl-CoA N-acyltransferase; Aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; Benzoyl-coenzyme A:glycine N-acyltransferase; Glycine N-acyltransferase; Glycine N-acyltransferase Acyl-CoA:glycine N-acyltransferase AAc Aralkyl acyl-CoA N-acyltransferase Aralkyl acyl-CoA:amino acid N-acyltransferase Benzoyl-coenzyme A:glycine N-acyltransferase Glycine N-benzoyltransferase HRP-1(CLP) GLYAT ACGNAT CAT GAT; Glycine N-benzoyltransferase; HRP-1(CLP)
Gene Aliases: ACGNAT; CAT; GAT; GLYAT
UniProt ID: (Human) Q6IB77
Entrez Gene ID: (Human) 10249
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.