Novus Biologicals
Manufacturer Code:NBP299587V
Catalog # NB061869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GLYAT. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAc; ACGNATAAc Acyl-CoA:glycine N-acyltransferase Aralkyl acyl-CoA N-acyltransferase Aralkyl acyl-CoA:amino acid N-acyltransferase aralkyl-CoA N-acyltransferase CATEC 2.3.1.13 GATHRP-1(CLP) glycine N-acyltransferase glycine-N-acyltransferase; Acyl-CoA:glycine N-acyltransferase; Aralkyl acyl-CoA N-acyltransferase; Aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; Benzoyl-coenzyme A:glycine N-acyltransferase; Glycine N-acyltransferase; Glycine N-benzoyltransferase; HRP-1(CLP)
Gene Aliases: ACGNAT; CAT; GAT; GLYAT
UniProt ID: (Human) Q6IB77
Entrez Gene ID: (Human) 10249
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.