Proteintech
Manufacturer Code:109001AP150UL
Catalog # 50-172-6696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLYAT Fusion Protein Ag1351 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAc; AAc ACGNAT CAT GAT GLYAT glycine N acyltransferase HRP 1(CLP); Acyl-CoA:glycine N-acyltransferase; Aralkyl acyl-CoA N-acyltransferase; Aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; Benzoyl-coenzyme A:glycine N-acyltransferase; Glycine N-acyltransferase; Glycine N-benzoyltransferase; HRP-1(CLP)
Gene Aliases: A330009E03Rik; ACGNAT; AI195249; AI315345; CAT; GAT; GLYAT
UniProt ID: (Human) Q6IB77, (Mouse) Q91XE0
Entrez Gene ID: (Human) 10249, (Mouse) 107146
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.