Novus Biologicals
Manufacturer Code:NBP247448
Catalog # NBP247448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: NRNSKELGLVPLTDDTSHARPPGPGRALLECDHLRSGVPGGRRRKDWS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: facilitated glucose transporter member 9 GLUT-9 GLUT9Glut9 GLUTX solute carrier family 2 (facilitated glucose transporter) member 9 UAQTL2 urate voltage-driven efflux transporter 1; Glucose transporter type 9; GLUT-9; human glucose transporter-like protein-9; solute carrier family 2 (facilitated glucose transporter), member 9; Solute carrier family 2, facilitated glucose transporter member 9; Urate transporter; urate voltage-driven efflux transporter 1
Gene Aliases: GLUT9; GLUTX; SLC2A9; UAQTL2; URATv1
UniProt ID: (Human) Q9NRM0
Entrez Gene ID: (Human) 56606
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.