Invitrogen
Manufacturer Code:PA580024
Catalog # PIPA580024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of mouse SLC2A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fructose transporter; Glucose transporter type 5, small intestine; GLUT-5; solute carrier family 2 (facilitated glucose transporter), member 5; Solute carrier family 2 facilitated glucose transporter member 5 Fructose transporter Glucose transporter type 5 small intestine GLUT-5 Slc2a5 Glut5; Solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5
Gene Aliases: AI526984; Glut5; Slc2a5; Slc5a
UniProt ID: (Mouse) Q9WV38, (Rat) P43427
Entrez Gene ID: (Mouse) 56485, (Rat) 65197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.