Invitrogen
Manufacturer Code:PA580023
Catalog # PIPA580023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human SLC2A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fructose transporter; Glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT-5; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; Solute carrier family 2 facilitated glucose transporter member 5 Fructose transporter Glucose transporter type 5 small intestine GLUT-5 SLC2A5 GLUT5; Solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81
Gene Aliases: GLUT-5; GLUT5; SLC2A5
UniProt ID: (Human) P22732, (Rat) P43427
Entrez Gene ID: (Human) 6518, (Rat) 65197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.