Invitrogen
Manufacturer Code:PA580022
Catalog # PIPA580022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human GLUT4 recombit protein (Position: N333-D509). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; Glucose transporter type 4, insulin-responsive; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter type 4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; Solute carrier family 2 facilitated glucose transporter member 4 Glucose transporter type 4 insulin-responsive GLUT-4 SLC2A4 GLUT4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: Glut-4; GLUT4; SLC2A4
UniProt ID: (Human) P14672, (Rat) P19357, (Mouse) P14142
Entrez Gene ID: (Human) 6517, (Rat) 25139, (Mouse) 20528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.