Invitrogen
Manufacturer Code:PA523052
Catalog # PIPA523052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human GLUT4 protein (between residues 480-509) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; Glucose transporter type 4, insulin-responsive; GLUT-4; GLUT4 GLUT-4 glucose transporter type 4 insulin-responsive insulin-responsive glucose transporter type 4 solute carrier family 2 facilitated glucose transporter member 4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter type 4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: Glut-4; GLUT4; SLC2A4
UniProt ID: (Human) P14672, (Rat) P19357, (Mouse) P14142
Entrez Gene ID: (Human) 6517, (Rat) 25139, (Mouse) 20528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.