Invitrogen
Manufacturer Code:PA11065
Catalog # APP-1065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues E(493) Q E V K P S T E L E Y L G P D E N D(509) of rat GLUT4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; Glucose transporter type 4 insulin-responsive; Glucose transporter type 4, insulin-responsive; GLUT-4; insulin responsive glucose transporter; solute carrier family 2 member 4; solute carrier family 2 , member 4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: Glut4; Slc2a4
UniProt ID: (Rat) P19357
Entrez Gene ID: (Rat) 25139
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.