Invitrogen
Manufacturer Code:720238
Catalog # PI720238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptides corresponding to Human GLUT2 (aa 73-86 264-281 498-511) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 2, liver; GLUT-2; liver-type glucose transporter; solute carrier family 2 (facilitated glucose transporter), member 2; Solute Carrier Family 2 Member 2; Solute carrier family 2, facilitated glucose transporter member 2
Gene Aliases: AI266973; Glut-2; GLUT2; GTT2; SLC2A2
UniProt ID: (Human) P11168, (Mouse) P14246
Entrez Gene ID: (Human) 6514, (Mouse) 20526, (Rat) 25351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.