Novus Biologicals
Manufacturer Code:NBP15968820UL
Catalog # NBP15968820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC2A10(solute carrier family 2 (facilitated glucose transporter) member 10) The peptide sequence was selected from the middle region of SLC2A10. Peptide sequence AKKTKPHPRSGDPSAPPRLALSSALPGPPLPARGHALLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATS Glucose transporter type 10 GLUT-10 GLUT10MGC126706 solute carrier family 2 (facilitated glucose transporter) member 10 solute carrier family 2 facilitated glucose transporter member 10; Glucose transporter type 10; GLUT-10; solute carrier family 2 (facilitated glucose transporter), member 10; Solute carrier family 2, facilitated glucose transporter member 10
Gene Aliases: ATS; GLUT10; SLC2A10
UniProt ID: (Human) O95528
Entrez Gene ID: (Human) 81031
Molecular Function: carbohydrate transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.