Invitrogen
Manufacturer Code:PA11063
Catalog # APP1063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues S(475) D K T P E E L F H P L G A D S Q V(492) of rat GLUT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 1 erythrocyte/brain HepG2 glucose transporter; Glucose transporter type 1, erythrocyte/brain; GLUT-1; GT1; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2 a 1 (facilitated glucose transporter) brain; Solute carrier family 2, facilitated glucose transporter member 1; solute carrier family 2, member 1
Gene Aliases: Glut-1; GLUT1; GLUTB; GTG1; Gtg3; RATGTG1; SLC2A1
UniProt ID: (Bovine) P27674, (Mouse) P17809, (Rat) P11167
Entrez Gene ID: (Bovine) 282356, (Mouse) 20525, (Rat) 24778
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.