Proteintech
Manufacturer Code:144621AP150UL
Catalog # 50-172-9469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLUD2 Fusion Protein Ag5748 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: creeper; GDH 2; GDH 2 GDH2 GLUD2 GLUDP1 glutamate dehydrogenase 2; GluD2; gluR delta-2 subunit; Glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; glutamate receptor delta-2 subunit; Glutamate receptor ionotropic, delta-2; hotfoot; lurcher; minisatellite 10ac detected by probe MMS10; testicular secretory protein Li 14
Gene Aliases: B230104L07Rik; cpr; GDH2; GLUD2; GLUDP1; GluRdelta2; Grid2; ho; Lc; Lc<J>; MMS10-AC; Ms10ac; nmf408; tpr
UniProt ID: (Human) P49448, (Mouse) Q61625, (Rat) Q63226
Entrez Gene ID: (Human) 2747, (Mouse) 14804, (Rat) 79220
Molecular Function: dehydrogenase oxidoreductase ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.