Novus Biologicals
Manufacturer Code:NBP271910AF700
Catalog # N271910A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A10 |
Immunogen | Full length recombinant protein of human GLTP (NP_057517) produced in HEK293T cells. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GLTP; Glycolipid transfer protein; truncated glycolipid transfer protein
Gene Aliases: GLTP
UniProt ID: (Human) Q53Z13
Entrez Gene ID: (Human) 51228
Molecular Function:
nucleic acid binding
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.