Proteintech
Manufacturer Code:108501AP150UL
Catalog # 50-172-6654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLTP Fusion Protein Ag1293 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLTP; GLTP glycolipid transfer protein; Glycolipid transfer protein; truncated glycolipid transfer protein
Gene Aliases: 1110001F24Rik; C76925; C77564; GLTP; RGD1564442
UniProt ID: (Human) Q53Z13, (Rat) B0BNM9, (Mouse) Q9JL62
Entrez Gene ID: (Human) 51228, (Rat) 288707, (Mouse) 56356
Molecular Function:
nucleic acid binding
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.