Invitrogen
Manufacturer Code:PA564369
Catalog # PIPA564369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GLT8D4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucoside xylosyltransferase 2; Glucoside xylosyltransferase 2 242n2 Glycosyltransferase 8 domain-containing protein 4 GXYLT2 GLT8D4; glycosyltransferase 8 domain containing 4; Glycosyltransferase 8 domain-containing protein 4
Gene Aliases: GLT8D4; GXYLT2
UniProt ID: (Human) A0PJZ3
Entrez Gene ID: (Human) 727936
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.