Invitrogen
Manufacturer Code:PA524656
Catalog # PIPA524656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 14-42 amino acids from the N-terminal region of human GL8D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycosyltransferase 8 domain-containing protein 2; glycosyltransferase 8 domain-containing protein 2 gycosyltransferase
Gene Aliases: GALA4A; GLT8D2; UNQ1901/PRO4347
UniProt ID: (Human) Q9H1C3
Entrez Gene ID: (Human) 83468
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.