Invitrogen
Manufacturer Code:PA5112428
Catalog # PIPA5112428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Glycosyltransferase 8 domain-containing protein 2 protein (25-349AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GALA4A GLT8D2 glycosyltransferase 8 domain containing 2 Glycosyltransferase 8 domain-containing protein 2 gycosyltransferase UNQ1901/PRO4347; Glycosyltransferase 8 domain-containing protein 2
Gene Aliases: 1110021D20Rik; GALA4A; GLT8D2; UNQ1901/PRO4347
UniProt ID: (Human) Q9H1C3, (Mouse) Q640P4
Entrez Gene ID: (Human) 83468, (Mouse) 74782
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.