Invitrogen
Manufacturer Code:PA532140
Catalog # PIPA532140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 6 and 309 of Human GLT8D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP139 glycosyltransferase 8 domain-containing protein 1 glycosyltransferase AD-017
Gene Aliases: 2410004H05Rik; 5430414N14Rik; AI450005; Da2-24; Glt8d1
UniProt ID: (Rat) Q6AYF6, (Mouse) Q6NSU3
Entrez Gene ID: (Rat) 306253, (Mouse) 76485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.