Invitrogen
Manufacturer Code:PA5112978
Catalog # PIPA5112978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Glycosyltransferase 8 domain-containing protein 1 protein (33-332AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410004H05Rik 5430414N14Rik AD-017 AI450005 Da2-24 GALA4A GLT8D1 glycosyltransferase 8 domain containing 1 Glycosyltransferase 8 domain-containing protein 1 glycosyltransferase AD-017 MSTP137 MSTP139 UNQ572/PRO1134; Glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017
Gene Aliases: 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; GALA4A; GLT8D1; MSTP137; MSTP139; UNQ572/PRO1134
UniProt ID: (Human) Q68CQ7, (Mouse) Q6NSU3
Entrez Gene ID: (Human) 55830, (Mouse) 76485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.