Novus Biologicals
Manufacturer Code:NBP2849840.1ML
Catalog # NB032913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human GLT8D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycosyltransferase 8 domain containing 1 MSTP139; Glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017
Gene Aliases: AD-017; GALA4A; GLT8D1; MSTP137; MSTP139; UNQ572/PRO1134
UniProt ID: (Human) Q68CQ7
Entrez Gene ID: (Human) 55830
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.