Invitrogen
Manufacturer Code:PA519706
Catalog # PIPA519706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 550 to the C-terminus of Rat Glutamate Transporter 1 (EAAT2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAAT2 GLT-1 excitatory amino acid transporter 2 excitotoxic amino acid transporter 2 glial high affinity glutamate transporter glutamate/aspartate transporter II sodium-dependent glutamate/aspartate transporter 2 solute carrier family 1 member 2 solute carrier family 1 member 2; EAAT2 variant mEAAT2/5UT5; Excitatory amino acid transporter 2; glial high affinity glutamate transporter; GLT-1; GLUT-R; glutamate transporter 1c; high affinity glucose transporter; Sodium-dependent glutamate/aspartate transporter 2; Solute carrier family 1 member 2; solute carrier family 1, member 2
Gene Aliases: 1700091C19Rik; 2900019G14Rik; AI159670; Eaat2; Glt; GLT-1; GLT1; MGLT1; Slc1a2
UniProt ID: (Mouse) P43006, (Rat) P31596
Entrez Gene ID: (Mouse) 20511, (Rat) 29482
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.