Invitrogen
Manufacturer Code:PA5115151
Catalog # PIPA5115151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EAAT2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700091C19Rik 2900019G14Rik AI159670 EAAT2 EAAT2 variant mEAAT2/5UT5 excitatory amino acid transporter 2 excitotoxic amino acid transporter 2 glial high affinity glutamate transporter Glt Glt1 Glt-1 GLT1a GLT1b GLT1c glutamate transporter 1c glutamate/aspartate transporter II GLUT-R HBGT high affinity glucose transporter MGLT1 OTTHUMP00000165209 OTTHUMP00000165210 SLC1A2 sodium-dependent glutamate/aspartate transporter 2 solute carrier family 1 (glial high affinity glutamate transporter) member 2 solute carrier family 1 member 2 solute carrier family 1 member 2; EAAT2 variant mEAAT2/5UT5; Excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; GLT-1; GLUT-R; glutamate transporter 1c; Glutamate/aspartate transporter II; high affinity glucose transporter; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; Solute carrier family 1 member 2; solute carrier family 1, member 2
Gene Aliases: 1700091C19Rik; 2900019G14Rik; AI159670; EAAT2; Glt; GLT-1; GLT1; HBGT; MGLT1; SLC1A2
UniProt ID: (Human) P43004, (Mouse) P43006, (Rat) P31596
Entrez Gene ID: (Human) 6506, (Mouse) 20511, (Rat) 29482
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.