OriGene
Manufacturer Code:TA809494
Catalog # 50-168-2834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C10 |
Immunogen | Full length human recombit protein of human GLS2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: breast cell glutaminase; GA GLS LGA hLGA; GLS; glutaminase 2 (liver, mitochondrial); glutaminase I; Glutaminase liver isoform, mitochondrial; L-glutaminase; L-glutamine amidohydrolase; phosphate-activated glutaminase; phosphate-dependent glutaminase
Gene Aliases: GA; GLS; GLS2; hLGA; LGA
UniProt ID: (Human) Q9UI32
Entrez Gene ID: (Human) 27165
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.