Invitrogen
Manufacturer Code:PA578475
Catalog # PIPA578475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Mouse GLS2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A330074B06Rik AI195532 GA GLS Lga glutaminase 2 (liver mitochondrial); breast cell glutaminase; GLS; glutaminase 2 (liver, mitochondrial); glutaminase I; Glutaminase liver isoform, mitochondrial; L-glutaminase; L-glutamine amidohydrolase; LGA; liver mitochondrial glutaminase; phosphate-activated glutaminase; phosphate-dependent glutaminase
Gene Aliases: A330074B06Rik; AI195532; GA; GLS; GLS2; hLGA; Kiaa4146; LGA
UniProt ID: (Human) Q9UI32, (Rat) P28492, (Mouse) Q571F8
Entrez Gene ID: (Human) 27165, (Rat) 192268, (Mouse) 216456
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.